siRNA Details
| Gene Symbol: LOC102467655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105003.1 | 1 | 42 | |||
| GenBank mRNA | AA393216.1 | - | 1 | 42 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102467655| Gene Symbol: LOC102467655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105003.1 | 1 | 42 | |||
| GenBank mRNA | AA393216.1 | - | 1 | 42 | ||