siRNA Details
| Gene Symbol: FER1L6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103547.1 | 3, 4 | 903 | |||
| GenBank mRNA | AK057332.1 | - | 3, 4 | 903 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000520031.1 | - | 3, 4 | 903 | Chr. 8 - 124046073 - 124171522 [-] on Build GRCh38 | |

FER1L6-AS2
Genomic Map