siRNA Details
| Gene Symbol: LOC101929488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125818.1 | 9 | 1799 | |||
| GenBank mRNA | BC039330.1 | - | 9 | 1799 | ||
| BC048118.1 | - | 9 | 1798 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519714.1 | - | 9 | 1799 | Chr. 8 - 57343787 - 57364857 [+] on Build GRCh38 | |

LOC101929488
Genomic Map