siRNA Details
Gene Symbol: LOC102724612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125825.1 | 2 | 1043 | |||
GenBank mRNA | BC040302.1 | - | 2 | 1044 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000518739.1 | - | 2 | 1043 | Chr. 8 - 63465849 - 63475482 [+] on Build GRCh38 |
