siRNA Details
| Gene Symbol: LINC01419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122034.1 | 2 | 671 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000522365.1 | - | 2 | 671 | Chr. 8 - 83403758 - 83408900 [+] on Build GRCh38 | |

LINC01419
Genomic Map