siRNA Details
Gene Symbol: CASC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117099.1 | 3 | 385 | |||
GenBank mRNA | AL709378.1 | - | 3 | 387 | ||
JX003867.1 | - | 1 | 90 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CASC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117099.1 | 3 | 385 | |||
GenBank mRNA | AL709378.1 | - | 3 | 387 | ||
JX003867.1 | - | 1 | 90 |