siRNA Details
Gene Symbol: LOC101926892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110653.1 | 4 | 1772 | |||
GenBank mRNA | AK024336.1 | - | 4 | 1767 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520259.1 | - | 4 | 1772 | Chr. 8 - 70471134 - 70485687 [+] on Build GRCh38 |
