siRNA Details
| Gene Symbol: NRG1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104156.1 | 1 | 52 | |||
| NR_104157.1 | 1 | 52 | ||||
| GenBank mRNA | BC037250.1 | - | 1 | 51 | ||
| BU567364.1 | - | 1 | 42 | |||
| HY002956.1 | - | 1 | 52 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000521463.5 | - | 1 | 51 | Chr. 8 - 32026219 - 32099765 [+] on Build GRCh38 | |

NRG1-IT1
Genomic Map