siRNA Details
| Gene Symbol: NRG1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037250.1 | - | 4 | 657 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000521463.5 | - | 4 | 657 | Chr. 8 - 32026219 - 32099765 [+] on Build GRCh38 | |

NRG1-IT1
Genomic Map