siRNA Details
| Gene Symbol: LOC101927752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134302.1 | 2 | 1141 | |||
| NR_134303.1 | 2 | 980 | ||||
| GenBank mRNA | BI860307.1 | - | 1 | 566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927752| Gene Symbol: LOC101927752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134302.1 | 2 | 1141 | |||
| NR_134303.1 | 2 | 980 | ||||
| GenBank mRNA | BI860307.1 | - | 1 | 566 | ||