siRNA Details
| Gene Symbol: LOC100507071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134293.1 | 2 | 266 | |||
| GenBank mRNA | BX099390.1 | - | 2 | 266 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000523556.1 | - | 2 | 266 | Chr. 8 - 22254576 - 22275162 [-] on Build GRCh38 | |

LOC100507071
Genomic Map