siRNA Details
Gene Symbol: LINC01299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033893.1 | 3 | 982 | |||
GenBank mRNA | AK094960.1 | - | 3 | 968 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520902.1 | - | 3 | 1096 | Chr. 8 - 65527008 - 65562780 [-] on Build GRCh38 |
