siRNA Details
| Gene Symbol: CASC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI223175.1 | - | 380 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC19| Gene Symbol: CASC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI223175.1 | - | 380 | |||