siRNA Details
| Gene Symbol: CASC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120364.1 | 2, 3 | 233 | |||
| GenBank mRNA | BI223175.1 | - | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC19| Gene Symbol: CASC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120364.1 | 2, 3 | 233 | |||
| GenBank mRNA | BI223175.1 | - | 456 | |||