siRNA Details
| Gene Symbol: LOC157273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040039.1 | 3 | 2763 | |||
| GenBank mRNA | AK055863.1 | - | 3 | 2763 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000520390.1 | - | 3 | 2763 | Chr. 8 - 9325051 - 9335080 [+] on Build GRCh38 | |

LOC157273
Genomic Map