siRNA Details
| Gene Symbol: LOC101929415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125813.1 | 1 | 303 | |||
| GenBank mRNA | AK091563.1 | - | 1 | 302 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518662.5 | - | 1 | 303 | Chr. 8 - 56445807 - 56552067 [+] on Build GRCh38 | |

LOC101929415
Genomic Map