siRNA Details
| Gene Symbol: LOC101929415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125813.1 | 4 | 2090 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518662.5 | - | 4 | 2090 | Chr. 8 - 56445807 - 56552067 [+] on Build GRCh38 | |

LOC101929415
Genomic Map