siRNA Details
Gene Symbol: LINC00824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121672.1 | 8 | 3006 | |||
GenBank mRNA | BC015157.1 | - | 1 | 1172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520206.5 | - | 6 | 2030 | Chr. 8 - 128405269 - 128427916 [-] on Build GRCh38 |
