siRNA Details
| Gene Symbol: LY96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013299.1 | XP_016868788.1 | 5 | 737 | ||
| XM_017013300.1 | XP_016868789.1 | 4 | 768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY96| Gene Symbol: LY96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013299.1 | XP_016868788.1 | 5 | 737 | ||
| XM_017013300.1 | XP_016868789.1 | 4 | 768 | |||