siRNA Details
| Gene Symbol: LOC101927815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125426.1 | 2 | 182 | |||
| GenBank mRNA | AK128880.1 | - | 2 | 148 | ||
| DA774838.1 | - | 2 | 120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927815| Gene Symbol: LOC101927815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125426.1 | 2 | 182 | |||
| GenBank mRNA | AK128880.1 | - | 2 | 148 | ||
| DA774838.1 | - | 2 | 120 | |||