siRNA Details
| Gene Symbol: LOC101929172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104677.1 | 3 | 708 | |||
| GenBank mRNA | AK057515.1 | - | 3 | 708 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519996.1 | - | 3 | 708 | Chr. 8 - 21298239 - 21309449 [+] on Build GRCh38 | |

LOC101929172
Genomic Map