siRNA Details
| Gene Symbol: LOC101928132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135052.1 | 2 | 621 | |||
| GenBank mRNA | BC015849.1 | - | 2 | 625 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000532065.1 | - | 2 | 621 | Chr. 11 - 13921450 - 13924863 [-] on Build GRCh38 | |

LOC101928132
Genomic Map