siRNA Details
Gene Symbol: LOC101928132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135052.1 | 2 | 476 | |||
GenBank mRNA | BC015849.1 | - | 2 | 480 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000532065.1 | - | 2 | 476 | Chr. 11 - 13921450 - 13924863 [-] on Build GRCh38 |
