siRNA Details
| Gene Symbol: LOC105376114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135839.1 | 2 | 638 | |||
| NR_135840.1 | 2 | 283 | ||||
| GenBank mRNA | AK300656.1 | - | 2 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376114| Gene Symbol: LOC105376114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135839.1 | 2 | 638 | |||
| NR_135840.1 | 2 | 283 | ||||
| GenBank mRNA | AK300656.1 | - | 2 | 275 | ||