siRNA Details
| Gene Symbol: LOC105376114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135839.1 | 2 | 1252 | |||
| NR_135840.1 | 2 | 897 | ||||
| GenBank mRNA | AK300656.1 | - | 2 | 889 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000531661.1 | - | 2 | 910 | Chr. 9 - 83708318 - 83713378 [+] on Build GRCh38 | |

LOC105376114
Genomic Map