siRNA Details
| Gene Symbol: LOC105376554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135109.1 | 2 | 1260 | |||
| GenBank mRNA | AK127718.1 | - | 2 | 1261 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000531559.1 | - | 2 | 1260 | Chr. 11 - 12030875 - 12061785 [+] on Build GRCh38 | |

LOC105376554
Genomic Map