siRNA Details
| Gene Symbol: ERVH-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U88898.1 | - | 3 | 771 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000527562.1 | - | 3 | 771 | Chr. Y - 21038289 - 21044724 [-] on Build GRCh38 | |

ERVH-6
Genomic Map