siRNA Details
Gene Symbol: LOC105376633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135110.1 | 1 | 52 | |||
GenBank mRNA | BX115137.1 | - | 1 | 53 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000532562.1 | - | 1 | 52 | Chr. 11 - 37938601 - 37954663 [+] on Build GRCh38 |
