siRNA Details
| Gene Symbol: LOC105376633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135110.1 | 1, 2 | 110 | |||
| GenBank mRNA | BX115137.1 | - | 1, 2 | 111 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000532562.1 | - | 1, 2 | 110 | Chr. 11 - 37938601 - 37954663 [+] on Build GRCh38 | |

LOC105376633
Genomic Map