siRNA Details
| Gene Symbol: BBOX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125766.1 | 2 | 464 | |||
| NR_125767.1 | 2 | 104 | ||||
| NR_125768.1 | 2 | 147 | ||||
| GenBank mRNA | DA376689.1 | - | 2 | 116 | ||
| DA865512.1 | - | 2 | 116 | |||
| DB016334.1 | - | 2 | 104 | |||

BBOX1-AS1
Genomic Map