siRNA Details
| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120527.1 | 2 | 259 | |||
| GenBank mRNA | BX099517.1 | - | 2 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506675| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120527.1 | 2 | 259 | |||
| GenBank mRNA | BX099517.1 | - | 2 | 223 | ||