siRNA Details
| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120526.1 | 1 | 183 | |||
| NR_120527.1 | 1 | 183 | ||||
| GenBank mRNA | BX099517.1 | - | 1 | 147 | ||
| DB473712.1 | - | 1 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506675| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120526.1 | 1 | 183 | |||
| NR_120527.1 | 1 | 183 | ||||
| GenBank mRNA | BX099517.1 | - | 1 | 147 | ||
| DB473712.1 | - | 1 | 183 | |||