siRNA Details
| Gene Symbol: LOC101927702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035195.2 | - | 2 | 2332 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000532515.1 | - | 2 | 2332 | Chr. 14 - 60240121 - 60245653 [-] on Build GRCh38 | |

LOC101927702
Genomic Map