siRNA Details
| Gene Symbol: LOC101929269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134308.1 | 2 | 615 | |||
| GenBank mRNA | BC038544.1 | - | 2 | 615 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000532453.1 | - | 2 | 615 | Chr. 8 - 11062647 - 11067089 [-] on Build GRCh38 | |

LOC101929269
Genomic Map