siRNA Details
| Gene Symbol: LOC283299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036678.1 | 1 | 262 | |||
| GenBank mRNA | AK091855.1 | - | 1 | 262 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000527847.5 | - | 1 | 262 | Chr. 11 - 7850751 - 7905955 [-] on Build GRCh38 | |

LOC283299
Genomic Map