siRNA Details
Gene Symbol: LOC338667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018652.1 | XP_016874141.1 | 9 | 2674 | ||
XM_017018653.1 | XP_016874142.1 | 6 | 1721 | |||
XM_017018654.1 | XP_016874143.1 | 5 | 1424 |
