siRNA Details
| Gene Symbol: LOC101928823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120559.1 | 1, 2 | 233 | |||
| NR_120560.1 | 1, 2 | 281 | ||||
| GenBank mRNA | BX091057.1 | - | 1, 2 | 281 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000529827.1 | - | 1, 2 | 281 | Chr. 11 - 112484345 - 112487836 [+] on Build GRCh38 | |

LOC101928823
Genomic Map