siRNA Details
| Gene Symbol: LOC100499484-C9ORF174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036527.1 | 14, 15 | 1402 | |||
| NR_036528.1 | 14, 15 | 1402 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100499484-C9ORF174| Gene Symbol: LOC100499484-C9ORF174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036527.1 | 14, 15 | 1402 | |||
| NR_036528.1 | 14, 15 | 1402 | ||||