siRNA Details
| Gene Symbol: LOC101928896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120566.1 | 4 | 524 | |||
| GenBank mRNA | BC042472.1 | - | 4 | 524 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928896| Gene Symbol: LOC101928896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120566.1 | 4 | 524 | |||
| GenBank mRNA | BC042472.1 | - | 4 | 524 | ||