siRNA Details
| Gene Symbol: STX17-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038853.1 | 3 | 432 | |||
| GenBank mRNA | BC030123.1 | - | 3 | 438 | ||
| BG460404.1 | - | 1 | 257 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000529965.2 | - | 3 | 432 | Chr. 9 - 99886326 - 99906601 [-] on Build GRCh38 | |

STX17-AS1
Genomic Map