siRNA Details
| Gene Symbol: LOC105369423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135090.1 | 1 | 383 | |||
| GenBank mRNA | BX098897.1 | - | 1 | 388 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000532606.1 | - | 1 | 383 | Chr. 11 - 87718354 - 87719411 [-] on Build GRCh38 | |

LOC105369423
Genomic Map