siRNA Details
| Gene Symbol: LOC399886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122071.1 | 5 | 2579 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000528390.1 | - | 5 | 2579 | Chr. 11 - 45371397 - 45388508 [+] on Build GRCh38 | |

LOC399886
Genomic Map