siRNA Details
| Gene Symbol: LOC399886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122071.1 | 1 | 777 | |||
| GenBank mRNA | AK123417.1 | - | 1 | 778 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000528390.1 | - | 1 | 777 | Chr. 11 - 45371397 - 45388508 [+] on Build GRCh38 | |

LOC399886
Genomic Map