siRNA Details
| Gene Symbol: LOC171391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126342.1 | 2, 3 | 232 | |||
| GenBank mRNA | BU156985.1 | - | 2, 3 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC171391| Gene Symbol: LOC171391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126342.1 | 2, 3 | 232 | |||
| GenBank mRNA | BU156985.1 | - | 2, 3 | 202 | ||