siRNA Details
Gene Symbol: LOC101928424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135053.1 | 2 | 344 | |||
GenBank mRNA | BX104024.1 | - | 2 | 330 | ||
CR741883.1 | - | 2 | 345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135053.1 | 2 | 344 | |||
GenBank mRNA | BX104024.1 | - | 2 | 330 | ||
CR741883.1 | - | 2 | 345 |