siRNA Details
| Gene Symbol: CASC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125366.1 | 1 | 419 | |||
| GenBank mRNA | BC027619.2 | - | 1 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC23| Gene Symbol: CASC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125366.1 | 1 | 419 | |||
| GenBank mRNA | BC027619.2 | - | 1 | 399 | ||