siRNA Details
| Gene Symbol: LOC101927120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110136.1 | 4 | 1212 | |||
| NR_110137.1 | 3 | 1092 | ||||
| GenBank mRNA | AK092135.1 | - | 3 | 1002 | ||
| AK096987.1 | - | 4 | 1212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927120| Gene Symbol: LOC101927120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110136.1 | 4 | 1212 | |||
| NR_110137.1 | 3 | 1092 | ||||
| GenBank mRNA | AK092135.1 | - | 3 | 1002 | ||
| AK096987.1 | - | 4 | 1212 | |||