siRNA Details
Gene Symbol: LOC101929089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120572.1 | 4 | 636 | |||
NR_120573.1 | 3 | 529 | ||||
NR_120575.1 | 3 | 549 | ||||
NR_120576.1 | 3 | 361 | ||||
GenBank mRNA | AI885123.1 | - | 3 | 304 |
