siRNA Details
| Gene Symbol: LOC101929549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110112.1 | 4 | 1135 | |||
| GenBank mRNA | BC043558.1 | - | 4 | 1135 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000527518.1 | - | 4 | 1135 | Chr. 12 - 4700417 - 4720102 [-] on Build GRCh38 | |

LOC101929549
Genomic Map