siRNA Details
| Gene Symbol: LOC101928008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120539.1 | 3 | 357 | |||
| GenBank mRNA | U80769.1 | - | 3 | 369 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000533659.1 | - | 3 | 356 | Chr. 11 - 9839143 - 9929263 [+] on Build GRCh38 | |

LOC101928008
Genomic Map